Learn More
Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop
Use Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop for general laboratory needs including plating and quick and accurate sample throughput.
Brand: Technical Service Consultants™ TS/4-C
This item is not returnable.
View return policy
Description
- Cadmium-free pigments
- Gamma Irradiation to achieve a SAL of 10-6
- Shelf Life: 3 years
- 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Single-use | |
Hard Crystal Polystyrene | |
Clear | |
1000/Pk. | |
Ideal for heavily contaminated or clinical samples |
1 μL | |
Sterile | |
Inoculating Loop | |
Singles in peel pouch |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur