missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Hardened Steel Grinding Balls
£3.10 - £26.95
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
10174492
|
RETSCH
05.368.0108 |
15 mm |
£4.15
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
10771613
|
RETSCH
05.368.0057 |
30 mm |
£17.68
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
10771423
|
RETSCH
05.368.0056 |
40 mm |
£26.95
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
10054881
|
RETSCH
05.368.0059 |
10 mm |
£3.10
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
Specifications
Grinding Balls | |
Hardened Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Hardened Steel Grinding Balls