missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Hardness (Hardicol) Test Kit

Based on a unique colorimetric method, this Palintest Hardicol test is robust, reliable, accurate, and easy to use.

Brand:  Palintest™ PM254

Code : 04

Additional Details : Weight : 0.23500kg


Product Code. 11913617

  • £111.58 / Pack of 50
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • X50 test kit/photom total hard 0-50
  • Hardness (Hardicol) test kit
  • Hardness, Photometer Reagents, 0 - 500 mg/L CaCO3
  • The Palintest Hardicol test is based on a unique colorimetric method 
  • The reagents are provided in tablet form and the test is carried out simply by adding the appropriate tablets to a sample of the water
Specifications

Specifications

50
0 to 500mg/L
Hardness (Hardicol) Test Kit
50 Pack
50 Tests
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • photometry total hardness kit

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see medical advice on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ammonium chloride, boric acid, lithium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers