Learn More
Sartorius Hardware Package for Automatic Draft Shield
Activation of Motorized Draft Shield for Sartorius Cubis™ II ultra hi-resolution high-capacity micro balances.
Brand: Sartorius QAPP1002
Additional Details : Weight : 0.01000kg
Description
- Activation of Motorized Draft Shield for Sartorius Cubis™ II Ultra Hi-Resolution High-Capacity Micro Balances
- Removable and easy-to-clean; designed for tool-free disassembly
- Material: glass
- Configurable on models with MCA or MCE Interface at time of purchase
- After purchase licensing only available for MCA models
Specifications
Cubis™ II High-Capacity Micro Balance | |
Software License |