missing translation for 'onlineSavingsMsg'
Learn More

Harmine, 98%, Thermo Scientific Chemicals

Product Code. 15811012
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym leucoharmine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, harmine, yageine, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, telepathine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterin, banisterine
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Melting Point 262.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4649
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 212.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Harmine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'