Learn More
Brady™ Harsh Environment Multipurpose Die-cut Opaque Polyester Labels for M6/M7 Printers
Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 173369
 View more versions of this product
Product Code. 17849044
  • £40.50 / Pack of 100
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • UL approved, CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -70°C to 110°C (-94°F to 230°F)
  • Made of B-423 polyester material
Specifications
Specifications
Not Autoclavable
M7-26-423
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
100 Labels
-94°F to +230°F
0.003 in.
1.25 x 2.75 in.
2.75 in.
31.75 mm
Halogen-free (DIN VDE 0472 Part 815)
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory and Inspection Labeling
No
Rectangle
-70°C to +110°C
0.076 mm
1.25 in.
69.85 mm