Learn More
Brady™ Harsh Environment Multipurpose Polyester Labels for M710 and BMP71
Harsh Environment Multipurpose Polyester Labels for use with Brady™ Printers M710 and BMP71.
Brand:  Brady™ 173400
 View more versions of this product
Product Code. 17888864
  • £35.60 / Pack of 250
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -70°C to +110°C (-94°F to +230°F)
  • Brady™ authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability
  • A ribbon is required to print this material
Specifications
Specifications
Halogen-free (DIN VDE 0472 Part 815)
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-70°C
Square
-70°C to +110°C
0.076 mm
1 in.
25.4 mm
Permanent Acrylic
White
Label
Single-side Printable
Labels
Barcode Labeling, Circuit Board Labeling, Component and Equipment Labeling, General Identification, Inventory and Inspection Labeling
1 Label
250 Labels
-94°F to +230°F
0.003 in.
25.4 x 25.4 mm
1 in.
25.4 mm