missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hastelloy C∣r gauze, 100 mesh woven from 0.102mm (0.004in) dia wire, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Hastelloy C™ gauze |
---|---|
Name Note | Woven from 0.102mm (0.004 in.) dia. wire |
MDL Number | MFCD00801065 |
Mesh Size | 100 Mesh |
Form | Wire Cloth |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hastelloy C™ gauze | |
MFCD00801065 | |
Wire Cloth |
Woven from 0.102mm (0.004 in.) dia. wire | |
100 Mesh |
Safety and Handling
GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P501c
H334-H350-H372
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only