missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hastelloy External Probe

For Use With: Isotemp Hotplates and Hotplate Stirrers

Brand:  Fisherbrand™ CIC0001467

Product Code. 15323548

  • £106.00 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Hastelloy PT1000 Temperature probe for use with Isotemp and Isotemp Advanced hotplates and hotplate stirrers

  • Hastelloy PT1000 Temperature probe
  • Temp. Range: 0 to 400°C
  • Chemical and corrosion resistant
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Isotemp Hotplates and Hotplate Stirrers
0°C to 400°C
Probe
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers