missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ HATU Coupling Reagent
This is a coupling reagent for use in peptide synthesis.
£778.00 - £2745.00
Specifications
Content And Storage | Store in refrigerator (2°C to 8°C). |
---|---|
For Use With (Application) | Peptide Synthesis |
Product Type | Peptide Synthesis Resin |
Shipping Condition | Wet Ice |
Form | Resin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15890913
|
Applied Biosystems™
GEN076523 |
25 g |
£778.00
25g |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920663
|
Applied Biosystems™
GEN076525 |
100 g |
£2745.00
100g |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Store in refrigerator (2°C to 8°C). | |
Peptide Synthesis Resin | |
Resin |
Peptide Synthesis | |
Wet Ice |