Learn More
Hazard Warning Label 'Flammable' Self-Adhesive GHS and CLP Compliant 20x20mm
Brand:  N/A F028 01 021
Code : Z1
Additional Details : Weight : 0.07200kg
Product Code. 12889758
  • £47.73 / Pack of 1000
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
All hazard warning diamonds are laminated and produced on a polypropylene material. The base material used conforms with B.S 5609:1986 which is perfect for transport by Road, Rail, Air and Sea. Hazard warning diamonds fully comply with the GHS, CLP, RID IMDG/IMO, ADN and ICAO/IATA requirements.
They have the following main properties:
• Excellent Weather Resistance.
• Excellent Chemical Resistance.
• Excellent Abrasion Resistance.
• Marine Grade Adhesive.