missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Substance Labels, 76 mm Core
Pre-printed, benchtop printer labels for CLP-GHS hazardous substances.
Brand: Brady™ 622284
Additional Details : Weight : 1.15000kg
Description
- Supplied with pre-printed red diamonds
- Ready for you to print the required warning, compliant to the latest regulations
Specifications
Not Autoclavable | |
Red on White | |
Polypropylene (B-7594) | |
Non-sterile | |
BradyPrinter i5100, i7100, BradyPrinter PR Plus, and IP™ Series Printers, THT 76 mm Core Printers | |
1 | |
Rectangle | |
0.079 mm | |
75 mm |
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
Hazard Communication Identification | |
2 Diamond Layout | |
1000 Labels | |
-20°C to +100°C | |
115.4 mm | |
110 mm |