missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Expandable Rack

The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.

Brand:  asecos™ 27838

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 17268450

  • £538.48 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Flat-pack delivery with assembly instructions
  • Galvanised grids as storage levels and 1 bottom collecting sump made of galvanised steel.
Specifications

Specifications

5.3 gal.
Steel
39.84 in.
39.84 in.
78.74 in.
17.20 in.
105.82 lb.
20 L
Drum Rack
1012.00 mm
1012.00 mm
2000.00 mm
437.00 mm
48 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers