missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Expandable Rack
Shop All asecos Products
Click to view available options
Capacity (Metric):
25 L
30 L
32 L
41 L
45 L
205 L
Unit Size:
Each
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Flat-pack delivery with assembly instructions
- Galvanised grids as storage levels and 1 bottom collecting sump made of galvanised steel.
Specifications
Specifications
| Capacity (English) | 11.9 gal. |
| Capacity (Metric) | 45 L |
| Material | Steel |
| Type | Drum Rack |
| Length (English) | 39.61 in. |
| Length (Metric) | 1006.00 mm |
| Width (English) | 39.61 in. |
| Width (Metric) | 1006.00 mm |
| Height (English) | 78.74 in. |
| Height (Metric) | 2000.00 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Hazardous Substances Expandable Rack >
Spot an opportunity for improvement?Share a Content Correction