missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Expandable Rack
The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.
£702.78 - £868.14
Specifications
Material | Steel |
---|---|
Type | Drum Rack |
Height (English) | 78.74 in. |
Height (Metric) | 2000.00 mm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16491405
|
asecos™
27830 |
20 L |
£702.78
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
17816446
|
asecos™
27834 |
25 L |
£807.72
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
17834625
|
asecos™
27836 |
40 L |
£868.14
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Flat-pack delivery with assembly instructions
- 4 painted sumps made of steel as storage levels
Specifications
Steel | |
Drum Rack | |
78.74 in. | |
2000.00 mm |