missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
Brand: asecos™ 27872
This item is not returnable.
View return policy
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Rack system can be extended individually
- Flat-pack delivery with assembly instructions
- Rack with galvanised shelf as storage levels
- Collecting sump
- 100 mm ground clearance for transport with pallet truck
Specifications
59.4 gal. | |
Steel | |
97.24 in. | |
97.24 in. | |
82.68 in. | |
32.13 in. | |
438.72 lb. |
225 L | |
Drum Rack | |
2470.00 mm | |
2470.00 mm | |
2100.00 mm | |
816.00 mm | |
199 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Hazardous Substances Rack > Capacity: 225 L, 199 kg
Spot an opportunity for improvement?Share a Content Correction