missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

The Hazardous Substances Rack provides storage of aggressive chemicals.

Brand:  asecos™ 27859

Additional Details : Weight : 90.00000kg

 View more versions of this product

Product Code. 17836466

  • £1008.06 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Varioius storage level options
  • Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis)
Specifications

Specifications

10.6 gal.
Steel
53.54 in.
53.54 in.
78.74 in.
25.08 in.
198.41 lb.
40 L
Drum Rack
1360.00 mm
1360.00 mm
2000.00 mm
637.00 mm
90 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers