missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.

Brand:  asecos™ 27867

 View more versions of this product

Product Code. 17886186

  • £1715.00 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Rack with galvanised shelf as storage levels
  • Collecting sump
  • 100 mm ground clearance for transport with pallet truck
Specifications

Specifications

Blue
225 L
Drum Rack
2470.00 mm
2470.00 mm
2100.00 mm
816.00 mm
217 kg
59.4 gal.
Steel
97.24 in.
97.24 in.
82.68 in.
32.13 in.
478.40 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers