missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

Product Code. p-200003653 Shop All asecos Products
25 L, Each
Click to view available options
Capacity (Metric):
25 L
40 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17866556

Brand: asecos™ 27833

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • 4 painted sumps made of steel as storage levels

Specifications

Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Material Steel
Type Drum Rack
Length (English) 53.54 in.
Length (Metric) 1360.00 mm
Width (English) 53.54 in.
Width (Metric) 1360.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 17.20 in.
Depth (Metric) 437.00 mm
Weight (English) 182.98 lb.
Weight (Metric) 83 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
asecos™ Hazardous Substances Rack >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.