missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Woodsafe™ Sharpsbin and Hazardous waste containers with yellow single lid

Click to view available options
Unit Size:
Each
Pack of 20
Pack of 30
Capacity (Metric):
6 L
12 L
50 L
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
- Lowers the use of Fossil-based plastic by 80%!
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be securely permanently closed for transportation to the final disposal
- My Green Labs ACT Label
- Follows same process as yellow bins and can be used to transport the same substances
- Natural color and smell from wood - No migration of substances
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- Stop burning plastic
Specifications
Specifications
| Certifications/Compliance | EN ISO 23907-1:2019. UN3291, ADR Approved |
| Capacity (English) | 6.157 gal |
| Color | Brown, Yellow |
| Dimensions (L x W x H) | 39.5 x 39.5 x 23.8 cm (15.6 x 15.6 x 9.4 in.) |
| Disposable | Single-use |
| Stackable | Yes |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Includes | Bin, Lid, and Label |
| For Use With (Application) | Safe disposal of sharps and potentially infectious waste or medical waste |
| Show More |
Lower climate impact and ACT label
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction