Learn More
New Pig™ HazMat Spill Kits
Product Code. p-8010871
Encompass_Preferred
Click to view available options
Capacity (Metric):
13.62 L
34.068 L
79.49 L
For Use With (Application):
Acids, Bases, Unknowns
Acids, Bases, Unknowns Fluid Absorption
Oils, Coolants, Solvents, Water
Unit Size:
Each
Product Code. 12903595
Brand: New Pig™ 64CRSC
Only null left
Add to basket
Add to basket
  • Features absorbents proven for rapid response
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives,such as 98% sulfuric acid and 30% sodium hydroxide
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Prepacked container provides easy access to contents
  • Temporary disposal bags with ties help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
Specifications
Capacity (Metric) 13.62 L
Material Polyethylene, Polypropylene
Type Spill Kit
For Use With (Application) Acids, Bases, Unknowns Fluid Absorption
Includes 1 External dia. 3 in., 46 in. L PIG™ HazMat Chemical Absorbent Sock, 1 External dia. 3 in., 10 ft. L PIG™ HazMat Chemical Absorbent Sock , 4 L x W: 20 x 15 in. PIG™ HazMat Mat Pad, 6 L x W: 6 x 2 in. Tamperproof Seal Label, 2 W x H: 18 x 30 in. Polyethylene Disposal Bags