missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Horst™ HBQ Model Very Flexible Heating Tape
£323.00
Specifications
Electrical Requirements | ∼230V Voltage |
---|---|
Model | HBQ |
Product Type | Very Flexible Heating Tape |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10087141
|
Horst™
20402 |
1 m |
£323.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Specifications
∼230V Voltage | |
HBQ | |
Very Flexible Heating Tape |