missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HBSS (10X), no calcium, no magnesium, no phenol red
HBSS (10X), no calcium, no magnesium, no phenol red
Brand: Gibco™ 14185045
Code : 50
Additional Details : Weight : 0.50000kg
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14185-052). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications
ISO 13485 standards, FDA | |
Hanks' Balanced Salt Solution, 500mL, Liquid, 10X Concentration, 5.8 to 6.1 pH, With Glucose, 36 Months Shelf Life, For Mammalian Cell Culture | |
Liquid | |
Glucose | |
Mammalian Cell Culture | |
500 mL |
15 to 30°C | |
10 X | |
Hanks' Balanced Salt Solution | |
No Calcium, No Magnesium, No Sodium Bicarbonate, No Phenol Red | |
5.8 to 6.1 | |
For in-vitro diagnostic use |