missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HBSS, calcium, magnesium
HBSS, calcium, magnesium
£5.89 - £99.25
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 15 to 30°C |
Concentration | 1 X |
Form | Liquid |
Product Type | Hanks' Balanced Salt Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550616
|
Gibco™
24020083 |
100 mL |
£5.89
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11560616
|
Gibco™
24020091 |
500 mL |
£12.61
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11580616
|
Gibco™
24020141 |
20 x 100 mL |
£91.00
2000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11570616
|
Gibco™
24020133 |
10 x 500 mL |
£99.25
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility (24020-117). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
1 X | |
Hanks' Balanced Salt Solution | |
Mammalian Cell Culture | |
For in-vitro diagnostic use |
15 to 30°C | |
Liquid | |
Glucose, Calcium, Magnesium, Sodium Bicarbonate, Phenol Red | |
7.0 to 7.4 |
Safety and Handling
missing translation for 'shelfLife' : 36 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title