missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HBSS, with calcium and magnesium; no phenol red
HBSS, with calcium and magnesium. No phenol red.
£18.03 - £133.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Form | Liquid |
Product Type | Hanks' Balanced Salt Solution |
With Additives | Calcium, Magnesium, Glucose, Sodium Bicarbonate |
Without Additives | No Phenol Red |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550456
|
Gibco™
14025050 |
500 mL |
£18.03
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11560456
|
Gibco™
14025100 |
10 x 500 mL |
£133.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14025-134). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
Hanks' Balanced Salt Solution | |
No Phenol Red | |
For in-vitro diagnostic use |
Liquid | |
Calcium, Magnesium, Glucose, Sodium Bicarbonate | |
Mammalian Cell Culture |
Safety and Handling
missing translation for 'shelfLife' : 36 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title