missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red
HBSS, no calcium, no magnesium, no phenol red
£7.19 - £156.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 15 to 30°C |
Concentration | 1 X |
Form | Liquid |
Product Type | Hanks' Balanced Salt Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520476
|
Gibco™
14175046 |
100 mL |
£7.19
100mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11530476
|
Gibco™
14175053 |
500 mL |
£18.02
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11550476
|
Gibco™
14175137 |
20 x 100 mL |
£117.00
2000mL |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11540476
|
Gibco™
14175129 |
10 x 500 mL |
£156.00
5000mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
1 X | |
Hanks' Balanced Salt Solution | |
No Calcium, No Magnesium, No Phenol Red | |
6.7 to 7.8 |
15 to 30°C | |
Liquid | |
Glucose, Sodium Bicarbonate | |
Mammalian Cell Culture | |
For in-vitro diagnostic use |