missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ HCoV-OC43 Spike Protein (YP_009555241.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3131
Additional Details : Weight : 0.00970kg
Specifications
SARS-CoV-2 Spike | |
10 μg at 0.2 μg/μL | |
4075 bp | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
HCoV-OC43 |
YP_009555241.1 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
43740568 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |