missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCS CellMask™ Stains
Klicka för att se tillgängliga alternativ
Description:
HCS CellMask Blue Stain
HCS CellMask Deep Red Stain
HCS CellMask Green Stain
HCS CellMask Near-IR Stain
HCS CellMask Orange Stain
HCS CellMask™ Red Stain
Förpackningsstorlek:
1 set
Each
Beskrivning
Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.
The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.
HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.
HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.
Beställningsinfo
Shipping Condition: Room temperature
Specifikationer
Specifikationer
| Color | Orange |
| Content And Storage | Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates. Store at ≤-20°C, desiccate, and protect from light. |
| Excitation Wavelength Range | 556/572 |
| For Use With (Equipment) | High Content Instrument |
| Quantity | 1 Set |
| Detection Method | Fluorescence |
| Description | HCS CellMask Orange Stain |
| Emission | Visible |
| Product Line | CellMask |
| Shipping Condition | Room Temperature |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering