missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS CellMask™ Stains

Product Code. 16481821
Click to view available options
Description:
HCS CellMask Blue Stain
HCS CellMask Deep Red Stain
HCS CellMask Green Stain
HCS CellMask Near-IR Stain
HCS CellMask Orange Stain
HCS CellMask™ Red Stain
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 16481821

Brand: Invitrogen™ H32722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easily visualize entire cells or individual cell structures during high-content screening (HCS) assays with HCS CellMask stains, which are available in a variety of colors for multiplexing flexibility and can be used immediately after fixation and permeabilization, or after antibody labeling.

Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.

The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.

HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates.

Store at ≤-20°C, desiccate, and protect from light.

Format Vial(s)
For Use With (Equipment) High Content Instrument
Quantity 1 Set
Detection Method Fluorescence
Description HCS CellMask Near-IR Stain
Product Line CellMask
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Stain
SubCellular Localization Cytoplasm, Cytosol, Nucleus
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.