Learn More
Molecular Probes™ HCS NuclearMask™ Stains
Description
Easily identify cell nuclei in high-content screening (HCS) assays with the versatile HCS NuclearMask Blue, Red, and Deep Red stains, which survive standard formaldehyde-based fixation and detergent-based permeabilization methods and can be applied to live and fixed cells. Because their dyes are easily discriminated from other fluorophores, the HCS NuclearMask stains can be used in multiparametric analyses. They can also label the entire cell (i.e., cytoplasm and nucleus) and provide an accurate backdrop against which to assess the features of interest.
If total cell staining is desired, HCS CellMask stains label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. HCS CellMask stains are available in a choice of colors for multiplexing flexibility. Sufficient volume quantities are provided to stain ten 96-well plates using an assay volume of 100 μL per well.
Specifications
Specifications
| Color | Blue |
| Content And Storage | Includes 65 μL of 2000X concentrate, providing sufficient material to stain a total of ten 96-well microplates. Store at 2–6°C and protect from light. |
| Excitation Wavelength Range | 350/451 |
| For Use With (Equipment) | High Content Instrument |
| Quantity | 65 μL |
| Detection Method | Fluorescence |
| Description | HCS NuclearMask™ Blue Stain |
| Emission | Visible |
| Product Line | NuclearMask |
| Shipping Condition | Room Temperature |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.