Learn More
HDGF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562788
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GDGDKKGNAE GSSDEEGKLV IDEPAKEKNE KGALKRRAGD LLEDSPKRPK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.Specifications
HDGF | |
Polyclonal | |
Unconjugated | |
HDGF | |
AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51858 | |
HDGF | |
Recombinant protein corresponding to Human HDGF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only