Learn More
Bel-Art™ Scienceware™ HDPE Biohazard Bags
Made of High Molecular Weight, High-Density (HMHD) polyethylene. Bel Art™ HDPE Biohazard Bags are bright red, autoclavable, and feature printed biohazard symbol and sterilization indicator patch.
Brand: Bel-Art™ Scienceware™ F13166-0000
50.55 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 02
Additional Details : Weight : 0.18000kg
Description
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Size allows the bag to be inverted over your hand to safely pick up waste
- For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid™ Benchtop Biohazard Bag Holder (H13193-1000) and Cover (F13193-0102)
- Steam autoclavable at 121°C (250°F)
- Available in a box of 100 or 1000
- Sold separately: Poxygrid Benchtop Biohazard Bag Holder (H13193-1000) and Cover (F13193-0102)
Specifications
Biohazard Bag | |
Red | |
38 μm | |
280 mm |
Autoclavable | |
HDPE | |
220 mm |