Learn More
SymLine Safety Waste Caps
GREENER_CHOICE
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
Brand:  SymLine 106480
265.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10820411
  • £344.00 / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safety Waste Caps are available with different thread sizes for attachment to the pipe system and with different numbers of hose connections for disposal of the liquid waste.
  • Optionally, Safety Waste Caps are available with electronic level sensors that indicate the degree of filling of the disposal container.
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
  • Specifications
    Specifications
    Black
    SymLine System
    HDPE-EC
    Safety Waste Cap
    Waste Disposal System
    Electronic fill level control for connection table assembly
    S60/61
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    One moment while we fetch your results.
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    SymLine Safety Waste Caps > Thread: S60/61, electronic level control, GL25(m)