Learn More
SymLine Safety Waste Caps
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
Brand: SymLine 306489
57.72 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Waste Disposal System | |
2 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm | |
GL45 |
SymLine System | |
PTFE | |
Safety Waste Cap |
Product Content Correction
Product Title
SymLine Safety Waste Caps > Thread: GL45, 2x PFA-fittings
Spot an opportunity for improvement?Share a Content Correction