missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE Combination Closure

Product Code. 10156880
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Each

Product Code. 10156880

Brand: Kautex™ 2000083700

Only null left
Add to basket
Add to basket

Includes screw-on dropping tip and screw-on lid for the dropping tip. Kautex™ HDPE Combination Closures are suitable for dispensing adhesives without leaking or drying.

  • Dropping tip is cut to desired drop size
  • Screw closure ensures dropping tip is covered, preventing leaks
  • Manufactured from chemcal resistant HDPE
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material HDPE
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Screw-on Dropping Tip with Lid
For Use With (Equipment) 10/25/50 mL Dropper Bottles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.