missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE Easy Closure
Specifications
Color | Black |
Includes | PTFE-insert |
Material | HDPE |
Product Type | Easy Closure |
For Use With (Equipment) | UN Cylinders |
Closure Size | 500 to 2500 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'