missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ HDPE-EC Safety Canister and Cap
58.08 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Use promo code "23290" to get your promotional price.
Click to view available options
Type:
Canister
Replacement Screw Cap
Description
- Made of very robust and electronically conductive HDPE, which is highly resistant to all solvents commonly used in laboratories
- Filling quantities from 10 L for the use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g., professional SymLine™ Disposal Systems)
- Thread types S50 is commonly used in the laboratory and are compatible with SCAT™ Safety Waste Caps range for the safe disposal of volatile hazardous substances
- Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
- Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
- Ideally suited for combination with SCAT™ Safety Waste Caps or the SymLine™ Disposal Systems
Specifications
For Use With (Application) | Flammable Liquid Waste Collection |
Autoclavable | Not Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Certifications/Compliance | UN-X Approval |
Color | Black |
Dimensions (L x W x H) | 200 x 320 x 240 mm |
For Use With (Equipment) | Safety Funnel |
Includes | Canister with Viewing Strip |
Material | HDPE |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'