missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HDPE-EC Safety Canister and Cap

Catalog Number p-7250585
58.08 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.


Click to view available options
Type:
Canister
Replacement Screw Cap
This item is not returnable. View return policy
This item is not returnable. View return policy

Electrically conductive safety canisters made of high-quality HDPE-EC material. Due to their grounding capability, flying sparks caused by electrical discharge can be prevented when disposing of flammable or explosive laboratory liquids.

  • Made of very robust and electronically conductive HDPE, which is highly resistant to all solvents commonly used in laboratories
  • Filling quantities from 10 L for the use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g., professional SymLine™ Disposal Systems)
  • Thread types S50 is commonly used in the laboratory and are compatible with SCAT™ Safety Waste Caps range for the safe disposal of volatile hazardous substances
  • Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
  • Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
  • Ideally suited for combination with SCAT™ Safety Waste Caps or the SymLine™ Disposal Systems
For Use With (Application) Flammable Liquid Waste Collection
Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Certifications/Compliance UN-X Approval
Color Black
Dimensions (L x W x H) 200 x 320 x 240 mm
For Use With (Equipment) Safety Funnel
Includes Canister with Viewing Strip
Material HDPE
Thread Size S50
Type Canister
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'