missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE,Wrench
Click to view available options
For Use With (Equipment):
For Use with 5/10/20/25/30 L Industrial Jerrycans
Description
- Wrench HDPE tool for opening and closing
Specifications
Color | White |
For Use With (Equipment) | For Use with 5/10/20/25/30 L Industrial Jerrycans |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Screw Closure with Dispensing Pump |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'