Learn More
Kautex™ HDPE 308/310 Series Narrow Neck Bottle
Use this Kautex™HDPE 308/310 Series Narrow Neck Bottle for packaging, sending samples, mailing and storage. Suitable for filling and transport of hazardous liquids, it features a recessed labeling field with sizes 1000 and 2500 mL. and is UN approved for liquids.
Brand: Kautex™ 2000084153
Additional Details : Weight : 0.16200kg
Description
- Optimized handle for safe and comfortable handling.
- Recessed labelling field with sizes 1000 and 2500 mL.
- GL 45 neck, three different types of closures available
- leakproof
- UN approved for liquids
Specifications
2.5 L | |
28.7 mm | |
HDPE | |
UN 3H1/X1.9/250 | |
282 mm | |
127 mm |
Natural | |
Packaging, Sending Samples, Mailing, Storage | |
Narrow | |
Narrow | |
Square Narrow Mouth Bottle |