Learn More
SCAT HDPE Safety Funnels With Hinged Lid
The SCAT safety funnel with hinged lid is designed to save space and is easy to handle. Thanks to the hinged lid as a closing mechanism, even very small quantities and viscous liquid waste can be discharged easily. The hinged lid can be closed after use, which avoids evaporation.
Brand: SCAT 107670
169.14 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The SCAT safety funnel with hinged lid is designed to save space and is easy to handle. Due to the hinged lid as a closing mechanism, even very small quantities and viscous liquid waste can be discharged easily. Der Klappdeckel kann nach der Nutzung geschlossen werden, was Ausdunstungen Die Verschraubungen sind frei drehbar - dies erleichtert das Aufschrauben des Trichters. Alle Trichter sind aus schwarzem PE-HD, elektrisch leitfähig und mit einem Erdungskabel versehen, zur sicheren Erdung des Trichters. The dirt sieve serves as additional splash protection.Specifications
HDPE | |
Thread size: S 60/61;;en; | |
1 Ea. |
Red | |
With hinged lid | |
Safety Funnel |