missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ LDPE Narrow Neck Round Bottle Series 301 with Stopper, Ecru

Use the versatile Kautex™ LDPE Narrow Neck Round Bottle with Stopper, Ecru, for item packaging, sending samples, and transporting, mailing and storage.

Brand:  Kautex™ 2000770503

 View more versions of this product

Product Code. 17858097

  • £802.00 / Pack of 850

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stoppers packed in polybag
  • Flexible
  • Good shape-retaining properties
  • Leak-proof
  • LPDE
  • Narrow Neck
  • Color: Ecru
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
18 mm
850
Bottle
Yes
86 mm
18 mm
Screw Cap
Ecru
LDPE
1 Ea.
1 Ea.
Round
Bottle
37.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.