missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE, Screw Closure for Carboy with Stop Cock
Click to view available options
Natural; With red stopcock, For use with 20/25/30 L jerrycans
Description
- For use with 20/25/30 L jerrycans
- HDPE
- Natural colour
- With stop cock, red
Specifications
Capacity (Metric) | 20 L |
Certifications/Compliance | DIN60 |
Color | Natural |
For Use With (Equipment) | For Use with 20/25/30 L Jerrycans |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Screw Closure with Stop Cock |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'