missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Screw Closure with stop cock (Natural)

Product Code. 10753111
Click to view available options
:
Natural; With red stopcock, For use with 2/2.5/3/6/8 L jerrycans
This item is not returnable. View return policy

Product Code. 10753111

Brand: Kautex™ 2000095872

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: 2/2.5/3/6/8 L jerrycans

  • Screw closure
  • HDPE
  • With red stop cock
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
For Use With (Equipment) For Use with 2/2.5/3/6/8 L Jerrycans
Material HDPE
No. per Pack 1 Ea.
Product Type Screw Closure
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.