missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Screw Closure for Venting System

Product Code. 10300271

Product Code. 10300271

Brand: Kautex™ 2000095912

Only null left
Add to basket
Add to basket

For Use With: 5 and 10 L jerrycans

  • Accommodates venting system for jerrycan
  • HDPE
  • Tamper-evident ring
  • Black
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) For Use with 5 and 10 L Jerrycans
Material HDPE
No. per Pack 1 Ea.
Product Type Screw Closure
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.