missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE, Screw Closure with Tamper-evident Ring (Black)
Click to view available options
Black; With tamper evident ring, For use with 5/10L jerrycans
Description
- Screw closure
- HDPE
- Tamper evident ring

Specifications
Specifications
Color | Black |
For Use With (Equipment) | For Use with 5/10 L Jerrycans |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Screw Closure |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Documents
Scientific Resources
Catalogs
Kautex - Expertise in packaging - Standard plastics
File size: 1.3 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kautex™ HDPE, Screw Closure with Tamper-evident Ring (Black) >
Spot an opportunity for improvement?Share a Content Correction