missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE, Screw Closure with Tamper-evident Ring (Red)
Click to view available options
Red; With tamper evident ring, For use with 20/25/30 L jerrycans
Description
- Screw closure
- HDPE
- Tamper evident ring
Specifications
Color | Red |
For Use With (Equipment) | For Use with 20/25/30 L Jerrycans |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Screw Closure |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'