missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Screw Closure with Tamper-evident Ring (Black)

Catalog Number 10098120
Click to view available options
Black; With tamper evident ring, For use with 20/25/30 L jerrycans

For Use With: 20/25/30 L jerrycans

Screw closure HDPE, black, with tamper evident ring
Color Black
For Use With (Equipment) For Use with 20/25/30 L Jerrycans
Material HDPE
No. per Pack 1 Ea.
Product Type Screw Closure
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'