missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE, Stopcock
Click to view available options
Material:
HDPE
Description
For use with Kautex™ Carboys
Specifications
Closure Type | Stopcock |
Color | Blue |
For Use With (Equipment) | For Carboy Series 350 |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Stopcock |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'