missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Universal Storage Containers

Practical storage containers with closed gripping edges for safe transport of pourable or liquid contents. Rounded inner edges and smooth inner surfaces for easycleaningenable use in the food stuffs/hygiene sector.

£50.00 - £120.00

Specifications

Certifications/Compliance Regulation (EC) No 1935/2004, Regulation (EU) 10/2011
Description Smooth inner finish
Color White
Material HDPE
Shape Rectangular
View More Specs

Alert:

Attention! Make sure to pay attention to the List of Chemical Resistance!


Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10040521
View Documents
Buerkle™
3418-0129
29 L
£50.00
Each
Please to purchase this item. Need a web account? Register with us today!
13117673
View Documents
Buerkle™
3418-0148
48 L
£70.95
Each
Please to purchase this item. Need a web account? Register with us today!
10434492
View Documents
Buerkle™
3418-0194
94 L
£120.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • HDPE, white
  • Can be stacked
Specifications

Specifications

Regulation (EC) No 1935/2004, Regulation (EU) 10/2011
White
Rectangular
Stackable
660 mm
Smooth inner finish
HDPE
Food Stuffs and Hygiene Sector
Storage Container
450 mm
Videos
SDS
Documents

Documents

Product Certifications